Diamond® PureFlow™ Nylon Syringe Filters, Non Sterile, Globe Scientific
Supplier: Globe Scientific
SF-NYLN-2230 SF-NYLN-4530 SF-NYLN-2213 SF-NYLN-4513
76516-636BX 272.65 CAD
76516-636 76516-638 76516-648 76516-650
Diamond® PureFlow™ Nylon Syringe Filters, Non Sterile, Globe Scientific
Filters Syringe Filters
Non sterile Diamond® PureFlow™ syringe filters are perfect for use in a wide range of filtration and sample applications where sterility is not needed. Non sterile filters are used in analytical chemistry for sample preperation, HPLC operations, and protecting analytical equipment from particle contamination.
  • Nylon membrane
  • High protein binding - not used when protein recovery is important
  • Aqueous or organic samples
  • Not compatible with acids, bases or for use in protein recovery
  • Ideal for general laboratory, chemical and beverage filtration and HPLC
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable