J&W VF-WAXms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP9205 CP9206 CP9207 CP9211 CP9202 CP9213 CP9214 CP9222 CP9223 CP9230
CAAGCP9205EA 1269.32 CAD
CAAGCP9205 CAAGCP9206 CAAGCP9207 CAAGCP9211 CAAGCP9202 CAAGCP9213 CAAGCP9214 CAAGCP9222 CAAGCP9223 CAAGCP9230
J&W VF-WAXms Columns, Agilent Technologies
Chromatography Columns
A high-performance polyethylene glycol PEG column for GC/MS food, flavor, and fragrance applications, especially where trace analyses are required.
  • Specially designed wax phase for accurate MS results with polar compounds
  • Operating temperature range of 20 to 250/260 °C
  • Improves signal-to-noise ratio for trace analysis
  • Improved performance with no change in the typical selectivity of PEG
  • 0.15 mm Int. Ø columns available for high efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SUPELCOWAX 10, SUPEROX II, CB-WAX, Stabilwax, BP-20, 007-CW, Carbowax, Rtx-WAX, ZB-WAX, ZB-WAX plus
These applications often require higher temperatures to analyze polar compounds, and therefore need an ultra-stable wax as a stationary phase. These very low bleed columns provide increased sensitivity, extended column lifetime, and greater accuracy, even at higher temperatures.