J&W VF-17ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP8986 CP5884 CP8981 CP8982 CP8984 CP9001
CACP8986EA 833.88 CAD
CACP8986 CAAGCP5884 CAAGCP8981 CAAGCP8982 CAAGCP8984 CAAGCP9001
J&W VF-17ms Columns, Agilent Technologies
Chromatography Columns
A 50% phenyl/50% dimethylpolysiloxane, medium-polarity, low-bleed column ideal for environmental and clinical research methods.
  • Ultralow bleed
  • Proprietary deactivation technology and manufacturing process improve column stability for enhanced column-to-column repeatability and column lifetime
  • Ideal for environmental and clinical methods
  • Ultralow bleed specification at 2 pA at 325 °C (0.25 mm × 30 m, 0.25 μm)
  • Ideal EPA confirmation column
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Low-bleed column that delivers increased sensitivity, accuracy, and enhanced mass spectral integrity. Often referenced in environmental and clinical research methods, this column features deactivation technology that improves column stability, resulting in improved repeatability and column lifetime.