Universal Bare Fused Silica Capillaries, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
160-2650-5 160-2644-5 160-2660-5 190-0131 190-0231 190-0331
CA160-2650-5EA 190.92 CAD
CA160-2650-5 CAAG160-2644-5 CAAG160-2660-5 CAAG190-0131 CAAG190-0231 CAAG190-0331
Universal Bare Fused Silica Capillaries, Agilent Technologies
Electrophoresis Systems Capillary Electrophoresis Systems
Fused silica 363 µm od capillaries with a window and 75 cm effective length, for any CE instrument.
  • To cut capillaries to the correct length, the CE column cutter is recommended (p/n 5183-4669)
Agilent provides 363 µm O.D, universal bare fused silica capillaries with a window and 75 cm effective length. These capillaries fit into any CE instrument.