J&W Ultra-2 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091B-101 19091B-102 19091B-102E 19091B-112 19091B-115 19091B-002 19091B-012 19091B-015 19091B-105 19091B-012E
CA19091B-101EA 886.55 CAD
CA19091B-101 CA19091B-102 CA19091B-102E CA19091B-112 CA19091B-115 CAAG19091B-002 CAAG19091B-012 CAAG19091B-015 CAAG19091B-105 CAAG19091B012E
J&W Ultra-2 Columns, Agilent Technologies
Chromatography Columns
Similar to HP-5 but with tighter specifications for retention index and capacity factors, for use with the Sadtler Retention Index Library.
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Ultra 2 is a (5%-phenyl)-methylpolysiloxane, nonpolar column that resembles HP-5, but with tighter specifications for retention index and capacity factors. It has an operating temperature range of –60 to 325/350 °C. These precision-engineered columns areMore Product Information