J&W Select Biodiesel Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP9078 CP9076
CAAGCP9078EA 1498.28 CAD
CAAGCP9078 CAAGCP9076
J&W Select Biodiesel Columns, Agilent Technologies
Chromatography Columns
Good column lifetime when operating at very high temperature up to 400 °C with ASTM and EN biodiesel methods.
  • Complete set of biodiesel columns for full compliance and ease-of-use
  • UltiMetal deactivation technology for stainless steel tubing provides high accuracy and longevity
  • Pre-tested for complete confidence in results
  • Convenient pre-coupled retention gap that is leak tested
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Although traditional fused silica can be used, high temperatures often mean shortened column lifetimes. Using an UltiMetal deactivated stainless steel column with an ultra-stable stationary phase produces more consistent results and virtually eliminates column breakage.