J&W PoraPLOT U Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7584 CP7581 CP7583
CAAGCP7584EA 1319.39 CAD
CAAGCP7584 CAAGCP7581 CAAGCP7583
J&W PoraPLOT U Columns, Agilent Technologies
Chromatography Columns
Provides excellent peak symmetry and retention time repeatability with polar and nonpolar volatiles.
  • Water has no effect on retention times and elutes as a sharp, quantifiable peak
  • Reliable retention time repeatability
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W PoraPLOT U provides symmetrical peaks for polar volatiles. Water has no effect on retention times and elutes as a sharp and quantifiable peak. PoraPLOT U is the most polar porous polymer PLOT column and is designed for halogenated compounds, hydrocarbons C1–C6, ketones, and solvents.