J&W PoraPLOT Q HT Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7557 CP7559
CAAGCP7557EA 1218.72 CAD
CAAGCP7557 CAAGCP7559
J&W PoraPLOT Q HT Columns, Agilent Technologies
Chromatography Columns
High-temperature version of PoraPLOT Q, offering the same benefits but operating up to 290 °C.
  • Recommended for column switching systems that analyze a broad range of polar and apolar volatile compounds
  • Long-term stability provides repeatable retention times
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rt-Q BOND, Rt-QPLOT, SupelQ PLOT
Agilent J&W PoraPLOT Q-HT is the high-temperature version of PoraPLOT Q, offering the same benefits for the analysis of polar and apolar volatile compounds but operating up to 290 °C. As with PoraPLOT Q, water elutes as a sharp and quantifiable peak, making retention times of target compounds repeatable, as retention is not influenced by water in the sample.