J&W PoraPLOT Q Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7549 CP7550 CP7551 CP7554 CP6954 CP7550I5 CP7552 CP7553
CAAGCP7549EA 1130.57 CAD
CAAGCP7549 CAAGCP7550 CAAGCP7551 CAAGCP7554 CAAGCP6954 CAAGCP7550I5 CAAGCP7552 CAAGCP7553
J&W PoraPLOT Q Columns, Agilent Technologies
Chromatography Columns
A porous polymer PLOT stationary phase for analysis of polar and apolar volatile compounds, and water as an analyte.
  • Long-term stability provides repeatable retention times
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rt-Q BOND, Rt-QPLOT, SupelQ PLOT
Agilent J&W PoraPLOT Q is recommended for column switching systems that analyze polar and apolar volatile compounds. Water elutes as a sharp and quantifiable peak, making retention times of target compounds repeatable, as they are not influenced by water in the sample.
Columns with 7" cage.