J&W PoraBOND U Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7381 CP7381I5
CAAGCP7381EA 1218.72 CAD
CAAGCP7381 CAAGCP7381I5
J&W PoraBOND U Columns, Agilent Technologies
Chromatography Columns
A highly stable polar-bonded porous polymer with a 300 °C maximum operating temperature.
  • Increased maximum temperature widens application range
  • Reduced bleed delivers lower detection limits and more accurate results
  • Bonded PLOT phase for longevity
  • Ideal for use with methods that utilize pressure programs or valve switching
  • Similar phases: Rt-U-BOND
The reduction of bleed provides lower detection limits and faster stabilization times. Because the porous polymer is bonded to the column, this column is ideal for use with pressure programs, GC/MSMore Product Information