J&W Porabond PoraBOND Q Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7352 CP7348 CP7350 CP7351 CP7353 CP7353I5 CP7354 CP7355
CAAGCP7352EA 1848.06 CAD
CAAGCP7352 CAAGCP7348 CAAGCP7350 CAAGCP7351 CAAGCP7353 CAAGCP7353I5 CAAGCP7354 CAAGCP7355
J&W Porabond PoraBOND Q Columns, Agilent Technologies
Chromatography Columns
An extremely stable bonded porous polymer PLOT stationary phase for analyzing volatile solvents and hydrocarbons.
  • Bonded PLOT column for more reliable results for analysis of volatile solvents and hydrocarbons
  • Extended analysis offers broad application range
  • Temprerature limits of 300 isothermal and 320 °C programmed
  • Engineered for high stability, withstands repeated water injections
  • Proprietary manufacturing technique results in very pure porous polymer with virtually no catalytic activity, allowing operation to 320 °C without decomposition
  • Bonding
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable