PLgel Individual Pore Size, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1110-6540 PL1110-6550 PL1110-6150 PL1110-6320 PL1110-8520 PL1510-5520
CAAGPL11106540EA 3628.68 CAD
CAAGPL11106540 CAAGPL11106550 CAAGPL11106150 CAAGPL11106320 CAAGPL11108520 CAAGPL15105520
PLgel Individual Pore Size, Agilent Technologies
Chromatography Columns
PLgel individual pore size columns offer high resolution over a mol wt range in organic solvents.
  • PLgel individual pore size GPC columns offer the highest resolution over a narrow molecular weight range
  • May be stacked to extend the molecular weight range
  • Recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns
PLgel individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. Columns may be stacked to extend the molecular weight range. Individual pore columns are generally recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns.