J&W DB-WAXetr Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-7332 122-7333 122-7362 123-7333 123-7334 125-7332 125-7334 122-7332E 122-7333E 122-7362E 122-7363 122-7332-INT 123-7312 123-7332 123-7354 123-7362 123-7363 123-7363E 123-7364 125-7312 125-7314 125-7332E 125-7333 125-7334E 125-7362 128-7323
CA122-7332EA 1143.36 CAD
CA122-7332 CA122-7333 CA122-7362 CA123-7333 CA123-7334 CA125-7332 CA125-7334 CAAG122-7332E CAAG122-7333E CAAG122-7362E CAAG122-7363 CAAG1227332INT CAAG123-7312 CAAG123-7332 CAAG123-7354 CAAG123-7362 CAAG123-7363 CAAG123-7363E CAAG123-7364 CAAG125-7312 CAAG125-7314 CAAG125-7332E CAAG125-7333 CAAG125-7334E CAAG125-7362 CAAG128-7323
J&W DB-WAXetr Columns, Agilent Technologies
Chromatography Columns
A high-polarity PEG column with an extended temperature range for alcohols, solvents, benzenes, styrenes, and organic acids.
  • Polyethylene glycol (PEG)
  • High polarity
  • Extended temperature range
  • Excellent column-to-column repeatability
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP phase G16
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SUPELCOWAX 10, SUPEROX II, CB-WAX, Stabilwax, BP-20, 007-CW, Carbowax, Rtx-WAX, ZB-WAX, ZB-WAX plus
Agilent J&W DB-WAXetr is an extended temperature range high-polarity polyethylene glycol (PEG) column for the analysis of alcohols, solvents, benzenes, styrenes, organic acids, and more. It has excellent column-to-column repeatability and is closely equivalent to USP phase G16.