J&W DB-23 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-2312 122-2332 122-2361 122-2362 122-2331 122-2332E 122-2362E 122-2332-INT 122-2361-INT 123-2332 123-2362 125-2312 125-2332
CA122-2312EA 674.94 CAD
CA122-2312 CA122-2332 CA122-2361 CA122-2362 CAAG122-2331 CAAG122-2332E CAAG122-2362E CAAG1222332INT CAAG1222361INT CAAG123-2332 CAAG123-2362 CAAG125-2312 CAAG125-2332
J&W DB-23 Columns, Agilent Technologies
Chromatography Columns
A high-polarity, bonded, and solvent-rinsable stationary phase for the separation of fatty acid methyl esters (FAMEs).
  • (50%-Cyanopropyl)-methylpolysiloxane
  • High polarity, replaces HP-23
  • Designed for separation of fatty acid methyl esters (FAMEs)
  • Excellent resolution for cis- and trans-FAME isomers
  • Close equivalent to USP phase G5
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-23 is specificallyMore Product Information