J&W CP-Volamine Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7447 CP7447I5 CP7448 CP7448I5
CACP7447EA 1610 CAD
CACP7447 CAAGCP7447I5 CAAGCP7448 CAAGCP7448I5
J&W CP-Volamine Columns, Agilent Technologies
Chromatography Columns
Optimized for the separation of volatile amines, even in the presence of water.
  • Nonpolar stationary phase
  • Excellent stability with water-containing samples expands the application range
  • Maximum temperature of 265 °C for enhanced productivity
  • High inertness provides sharp amine peaks for accurate results
  • Produces symmetrical peaks due to MPD (Multi-Purpose Deactivation) technology
  • Excellent performance even when the sample contains water (not as sample matrix)
  • Ideal for analyzing volatile amines like MMA, DMA, and TMA (monomethyl, dimethyl, and trimethyl amine)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-Volatile Amines
Agilent J&W CP-Volamine is optimized for the separation of volatile amines like MMA, DMA and TMA (monomethyl, dimethyl and trimethyl amine). The column is coated with a nonpolar stationary phase and produces symmetrical peaks with its Multi-Purpose Deactivation technology. It is the most stable column for analyzing volatile amines, even when the samples contain a high percentage of water.