J&W HP-PLOT Molesieve Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19095P-MS0 19095P-MS6 19091P-MS4 19091P-MS7 19091P-MS8 19091P-MS4E 19095P-MS5 19095P-MS9 19095P-MS0E 19095P-MS6E
CA19095P-MS0EA 2149.1 CAD
CA19095P-MS0 CA19095P-MS6 CAAG19091P-MS4 CAAG19091P-MS7 CAAG19091P-MS8 CAAG19091PMS4E CAAG19095P-MS5 CAAG19095P-MS9 CAAG19095PMS0E CAAG19095PMS6E
J&W HP-PLOT Molesieve Columns, Agilent Technologies
Chromatography Columns
Designed for the analysis of permanent gases.
  • A PLOT column for the analysis of permanent gases
  • O₂, N₂, CO, and CH₄ resolve in less than 5 minutes
  • Durable molecular sieve 5Å coating minimizes baseline spiking and damage to multiport valves
  • Select a thick film for Ar/O₂ separation without cryogenic cooling
  • Select thin film HP-PLOT Molesieve columns for routine air monitoring applications
  • Replaces GS-Molesieve
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each GC column