J&W HP-FFAP Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091F-102 19091F-112 19091F-413 19091F-433 19095F-120 19095F-123 19091F-105 19091F-115 19091F-102E 19091F-112E 19091F-115E 19095F-121 19095F-123E
CA19091F-102EA 983.68 CAD
CA19091F-102 CA19091F-112 CA19091F-413 CA19091F-433 CA19095F-120 CA19095F-123 CAAG19091F-105 CAAG19091F-115 CAAG19091F102E CAAG19091F112E CAAG19091F115E CAAG19095F-121 CAAG19095F123E
J&W HP-FFAP Columns, Agilent Technologies
Chromatography Columns
HP-FFAP is a high-polarity phase designed for the analysis of volatile fatty acids and phenols.
  • Nitroterephthalic acid modified polyethylene glycol
  • Designed for the analysis of volatile fatty acids and phenols
  • Replaces OV-351
  • Bonded and cross-linked, solvent rinsable
  • Close equivalent to USP Phase G35
  • Similar Phases: Stabilwax-DA, Nukol, 007-FFAP, BP21, AT-1000, OV-351
Agilent J&W HP-FFAP is designed primarily for the analysis of organic acids, free fatty acids, or samples that require quantitation of acidic impurities. The FFAP stationary phase isMore Product Information