HP-5 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091J-012 19091J-101 19091J-215 19091J-413E 19091J-431 19091J-433 19095J-121 19095J-321 19095J-623 19091J-202 19091J-413 19091J-005 19091J-102 19091J-112 19091J-113 19091J-115 19091J-205 19091J-212 19091J-213 19091J-216 19091J-231 19091J-233 19091J-236 19091J-333 19091J-411 19091J-416 19091J-436 19091J-577 19091J-012E 19091J-112E 19091J-215E 19091J-216E 19091J-411E 19091J-413-INT 19091J-433E 19091J-577LTM 19095J-023 19095J-123 19095J-323 19095J-621
CA19091J-012EA 1063.52 CAD
CA19091J-012 CA19091J-101 CA19091J-215 CA19091J-413E CA19091J-431 CA19091J-433 CA19095J-121 CA19095J-321 CA19095J-623 CA19091J-202 CA76514-570 CAAG19091J-005 CAAG19091J-102 CAAG19091J-112 CAAG19091J-113 CAAG19091J-115 CAAG19091J-205 CAAG19091J-212 CAAG19091J-213 CAAG19091J-216 CAAG19091J-231 CAAG19091J-233 CAAG19091J-236 CAAG19091J-333 CAAG19091J-411 CAAG19091J-416 CAAG19091J-436 CAAG19091J-577 CAAG19091J012E CAAG19091J112E CAAG19091J215E CAAG19091J216E CAAG19091J411E CAAG19091J413I CAAG19091J433E CAAG19091J577L CAAG19095J-023 CAAG19095J-123 CAAG19095J-323 CAAG19095J-621
HP-5 Columns, Agilent Technologies
Chromatography Columns
An excellent nonpolar, general-purpose 5% phenyl GC column with high temperature limits for a wide range of applications.
  • (5%-Phenyl)-methylpolysiloxane
  • An excellent nonpolar, general-purpose column for a wide range of applications
  • High temperature limit
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G27
  • Wide range of column dimensions available
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-5 is a precision-engineered (5%-phenyl)-methylpolysiloxane nonpolar column that delivers superb performance in a wide range of applications. This general-purpose column is bonded, crosslinked, and solvent rinsible and is equivalent to USP phase G27.