InfinityLab Poroshell 120 HILIC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
685775-601 823750-948 821725-947 673775-924 675775-924 679775-924 680970-924 683675-924 683775-924 683970-924 683975-324 683975-601 683975-924 685675-924 685775-924 685975-324 685975-924 689675-924 689775-601 689775-924 689975-324 689975-601 689975-924 820750-933
CAAG685775-601EA 2251.51 CAD
CAAG685775-601 CAAG823750-948 CA821725-947 CAAG673775-924 CAAG675775-924 CAAG679775-924 CAAG680970-924 CAAG683675-924 CAAG683775-924 CAAG683970-924 CAAG683975-324 CAAG683975-601 CAAG683975-924 CAAG685675-924 CAAG685775-924 CAAG685975-324 CAAG685975-924 CAAG689675-924 CAAG689775-601 CAAG689775-924 CAAG689975-324 CAAG689975-601 CAAG689975-924 CAAG820750-933
InfinityLab Poroshell 120 HILIC, Agilent Technologies
Chromatography Columns
Superficially porous particles for high-resolution HILIC separations with short equilibration times.
  • Superficially porous, 'core-shell' particles that provide improved throughput, shorter equilibration times, and higher resolution
  • A scalable family of particles—1.9, 2.7, and 4 μm—enable you to get the best from your methods and instruments
  • Separate many polar compound classes by choosing from orthogonal bare silica (HILIC), poly-hydroxy fructan (HILIC-OH5), or zwitterionic (HILIC-Z) phases
  • Fully compatible with Agilent InfinityLab deactivator additive,
More Product Information