J&W HP-PLOT Al2O3 S Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091P-S12 19091P-S15 19091P-S33 19095P-S25 19095P-S25E 19091P-S12PT 19095P-S21 19095P-S23 19095P-S23PT
CA19091P-S12EA 1343.75 CAD
CA19091P-S12 CA19091P-S15 CA19091P-S33 CA19095P-S25 CA19095P-S25E CAAG19091PS12P CAAG19095P-S21 CAAG19095P-S23 CAAG19095PS23P
J&W HP-PLOT Al2O3 S Columns, Agilent Technologies
Chromatography Columns
A deactivated, mid-polarity PLOT column for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers.
  • Middle range of polarity for alumina phases
  • Aluminum oxide deactivated with sodium sulfate
  • Excellent general-use column for light hydrocarbon analysis (C1–C8 isomers)
  • Best for resolving acetylene from butane and propylene from isobutane
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-PLOT Al₂O₃ S is the standard column choice for light hydrocarbon analysis, particularly C1 to C8 hydrocarbon isomers. A mid-polar range alumina phase deactivated with sodium sulfate, it is excellent for resolving acetylene from butylene and isobutane.