GS-GasPro Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
113-4312 113-4332 113-4362
CA113-4312EA 1381.73 CAD
CA113-4312 CA113-4332 CA113-4362
GS-GasPro Columns, Agilent Technologies
Chromatography Columns
A unique bonded silica PLOT column that is an excellent choice for light hydrocarbons and sulfur gases.
  • Unique bonded PLOT column technology
  • Excellent choice for light hydrocarbons and sulfur gases
  • Retention stability not affected by water
  • Separates CO and CO₂ on a single column
  • No particles, making it an ideal PLOT column for GC/MS
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: CP-Silica PLOT
The Agilent J&W GS-GasPro column is precision-engineered with unique bonded silica PLOT column technology, making it an excellent choice for light hydrocarbons and sulfur gases. With an operating temperature range of –80 to 260/300 °C and extensive testing to strict industry QC standards, it delivers the level of performance demanded by analytical GC/MS.
The column features high retention stability that is unaffected by water and enables separation of CO and CO2 on a single column.