DB-FFAP Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-3232 122-3233 123-3232 123-3232E 123-3234 125-3232 127-32H2 122-3212 122-3232E 122-3262 122-3263 123-3212 123-3223 123-3233 123-3233E 123-3253 123-3262 123-3263 123-3264 124-3232 125-3212 125-3217 125-3231 125-3233 125-3237 125-3262 127-3212
CA122-3232EA 1170.51 CAD
CA122-3232 CA122-3233 CA123-3232 CA123-3232E CA123-3234 CA125-3232 CAAG127-32H2 CAAG122-3212 CAAG122-3232E CAAG122-3262 CAAG122-3263 CAAG123-3212 CAAG123-3223 CAAG123-3233 CAAG123-3233E CAAG123-3253 CAAG123-3262 CAAG123-3263 CAAG123-3264 CAAG124-3232 CAAG125-3212 CAAG125-3217 CAAG125-3231 CAAG125-3233 CAAG125-3237 CAAG125-3262 CAAG127-3212
DB-FFAP Columns, Agilent Technologies
Chromatography Columns
DB-FFAP is designed for the analysis of volatile fatty acids and phenols.
  • Nitroterephthalic acid modified polyethylene glycol
  • High polarity
  • Designed for the analysis of volatile fatty acids and phenols
  • Bonded and cross-linked
  • solvent rinsable
  • Close equivalent to USP Phase G35
  • Replaces OV-351
  • Agilent J&W GC columns are tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency to give you utmost confidence in qualitative and quantitative results
  • For QC test conditions, including the test probe mixture and the
More Product Information