DB-ALC1 and DB-ALC2 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
123-9134 123-9234 125-9134 123-9134E 125-9234
CA123-9134EA 1197.28 CAD
CA123-9134 CA123-9234 CA125-9134 CAAG123-9134E CAAG125-9234
DB-ALC1 and DB-ALC2 Columns, Agilent Technologies
Chromatography Columns
Delivers blood alcohol analysis results in just 2 minutes, reducing run time by a third compared to other blood alcohol analysis columns.
  • Delivers reliable blood alcohol analysis
  • Optimized primary and confirmation column pair for US blood alcohol analysis
  • Faster GC run times
  • Improved resolution of key ethanol/acetone peaks
  • Available in 0.32 and 0.53 mm Int.Ø.
  • Bonded and crosslinked
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-ALC1 and DB-ALC2 analytical columns enable two-minute blood alcohol content analysis, reducing run time by one-third compared to other blood alcohol analysis columns. Because of its unique polarity, DB-ALC2 also provides better resolution of key ethanol/acetone peaks than other available blood alcohol confirmation columns.
These columns are suitable for both headspace analysis and direct injections.