DB-5ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
121-5522 122-5513 122-5516 122-5531 122-5533 122-5536 122-5561 122-5562 123-5532 123-5533 123-5536 123-5563 125-5532 128-5512 128-5522 122-5512 122-5532 122-5532E 122-5511 122-5522 122-5533E 122-5552 122-5562E 122-5563 122-5512LTM 122-5516-INT 122-5532-INT 123-5511 123-5512 123-5513 123-5526 123-5531 123-5532E 123-5562 123-5566 125-5512 125-5537 125-553J 128-5552
CA121-5522EA 1068.93 CAD
CA121-5522 CA122-5513 CA122-5516 CA122-5531 CA122-5533 CA122-5536 CA122-5561 CA122-5562 CA123-5532 CA123-5533 CA123-5536 CA123-5563 CA125-5532 CA128-5512 CA128-5522 CA76514-542 CA76514-546 CAAG122-5532E CAAG122-5511 CAAG122-5522 CAAG122-5533E CAAG122-5552 CAAG122-5562E CAAG122-5563 CAAG1225512LTM CAAG1225516INT CAAG1225532INT CAAG123-5511 CAAG123-5512 CAAG123-5513 CAAG123-5526 CAAG123-5531 CAAG123-5532E CAAG123-5562 CAAG123-5566 CAAG125-5512 CAAG125-5537 CAAG125-553J CAAG128-5552
DB-5ms Columns, Agilent Technologies
Chromatography Columns
A nonpolar column with high inertness and improved signal-to-noise for analysis of aromatics and semi volatiles.
  • Phenyl arylene polymer virtually equivalent to (5%-phenyl)-methylpolysiloxane
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Excellent inertness for active compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsible
  • Exact replacement of HP-5TA
  • Close equivalent to USP phase G27
  • Test mix available
  • Tested with the tightest industry QC specifications for column
More Product Information