J&W DB-35 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
123-1932 122-1932 123-1933 123-1933E 123-1968 125-1932 125-1937
CA123-1932EA 1197.71 CAD
CA123-1932 CAAG122-1932 CAAG123-1933 CAAG123-1933E CAAG123-1968 CAAG125-1932 CAAG125-1937
J&W DB-35 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column excellent for environmental analyses.
  • (35%-Phenyl)-methylpolysiloxane
  • Mid polarity - slightly more polar than HP-35
  • Inert to active solutes
  • Ideal for confirmational analyses
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP Phase G42
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-35 columns are ideal for conformational analysis. The DB-35 is a (35%-phenyl)-methylpolysiloxane, mid-polarity column. Bonded and crosslinked, it is inert to active solutes. DB-35 is a close equivalent to USP phase G42.