J&W DB-17ht Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-1831 122-1801 122-1811 123-1831
CA122-1831EA 1345.6 CAD
CA122-1831 CAAG122-1801 CAAG122-1811 CAAG123-1831
J&W DB-17ht Columns, Agilent Technologies
Chromatography Columns
A high-temperature GC column for improved resolution of triglycerides, ideal for confirmation analysis at elevated temperature.
  • (50%-Phenyl)-methylpolysiloxane
  • Mid-polarity
  • Extended upper temperature limit of 365 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
  • Improved resolution for triglycerides
  • Ideal for confirmation analysis
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance
More Product Information