DB-1701P Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-7732 125-7732 123-7732
CA122-7732EA 1218.05 CAD
CA122-7732 CA125-7732 CAAG123-7732
DB-1701P Columns, Agilent Technologies
Chromatography Columns
Bonded, crosslinked, and solvent rinsible, well suited for analysis of organochlorine pesticides.
  • Low/mid-polarity
  • Exact replacement of HP-PAS1701
  • Specifically designed and processed for the analysis of organochlorine pesticides
  • ECD tested to ensure minimal pesticide breakdown and low ECD bleed
  • Bonded, crosslinked, and solvent rinsible
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each GC column
Agilent J&W DB-1701P is specifically designed and processed for the analysis of organochlorine pesticides. These low- to mid-polarity columns are ECD tested to ensure minimal pesticide breakdown and low ECD bleed, and operate effectively within temperature limits of –20 to 260/300 °C.
These precision-engineered columns are tested to Agilent's stringent industry QC standards. As DB-1701P columns are bonded and crosslinked, they are also solvent rinsible and deliver high levels of accuracy and stability in a variety of applications.