J&W DB-1701 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-0732 122-0733 122-0762 123-0712 123-0732 123-0733 123-0763 125-0712 125-0732 121-0722 123-0713 123-0731 123-0732E 123-0733E 123-0753 123-0762 123-0733-INT 125-0731 125-0733 125-0762 127-0722 127-0723
CA122-0732EA 1154.47 CAD
CA122-0732 CA122-0733 CA122-0762 CA123-0712 CA123-0732 CA123-0733 CA123-0763 CA125-0712 CA125-0732 CAAG121-0722 CAAG123-0713 CAAG123-0731 CAAG123-0732E CAAG123-0733E CAAG123-0753 CAAG123-0762 CAAG1230733INT CAAG125-0731 CAAG125-0733 CAAG125-0762 CAAG127-0722 CAAG127-0723
J&W DB-1701 Columns, Agilent Technologies
Chromatography Columns
A low/mid polarity column ideal for the analysis of pesticides, herbicides, TMS sugars, and aroclors.
  • (14%-Cyanopropyl-phenyl)-methylpolysiloxane
  • Solvent rinsable with low/mid polarity
  • Bonded and crosslinked
  • Exact replacement of HP-1301 and HP-1701
  • Available with DuraGuard (the GC column with a built-in guard column)
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-1701, Rtx-1701, BP-10, OV-1701, 007-1701, ZB-1701
Agilent J&W DB-170More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Mid-polarity stationary phase
USP code G46