DB-17 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-1712 122-1732 123-1732 125-1732 122-1713 122-1731 122-1732E 122-1733 122-1762 122-1732LTM 123-1711 123-1712 123-1713 123-1733 123-1762 123-1732-INT 125-1704 125-1712 125-1713 125-1717 125-1731 125-1732E 125-1733 125-1762 127-1712 127-1722
CA122-1712EA 663.59 CAD
CA122-1712 CA122-1732 CA123-1732 CA125-1732 CAAG122-1713 CAAG122-1731 CAAG122-1732E CAAG122-1733 CAAG122-1762 CAAG1221732LTM CAAG123-1711 CAAG123-1712 CAAG123-1713 CAAG123-1733 CAAG123-1762 CAAG1231732INT CAAG125-1704 CAAG125-1712 CAAG125-1713 CAAG125-1717 CAAG125-1731 CAAG125-1732E CAAG125-1733 CAAG125-1762 CAAG127-1712 CAAG127-1722
DB-17 Columns, Agilent Technologies
Chromatography Columns
A mid-polarity column that is excellent for confirmation analysis.
  • (50%-Phenyl)-methylpolysiloxane
  • Mid polarity – slightly more polar than HP-50+
  • Excellent for confirmation analysis
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G3
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Rtx-50, 007-17(MPS-50), SP-2250, SPB-50, ZB-50, AT-50
Agilent J&W DB-17 is a (50%-phenyl)-methylpolysiloxane column of mid polarity. EquivalentMore Product Information