J&W DB-1301 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
123-1333 122-1332 122-1333 122-1363 123-1332 123-1363 125-1312 125-1332 125-1333
CA123-1333EA 1215.19 CAD
CA123-1333 CAAG122-1332 CAAG122-1333 CAAG122-1363 CAAG123-1332 CAAG123-1363 CAAG125-1312 CAAG125-1332 CAAG125-1333
J&W DB-1301 Columns, Agilent Technologies
Chromatography Columns
A low- to mid-polarity column, equivalent to USP Phase G43.
  • (6%-Cyanopropyl-phenyl)-methylpolysiloxane
  • Low- to mid-polarity
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-1301
  • Equivalent to USP Phase G43
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1301 (6% cyanopropyl-phenyl)-methylpolysiloxane column is a low- to mid-polarity, bonded, crosslinked, and solvent rinsable column, equivalent to USP phase G43.