J&W CP-Sil 8 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7595 CP7596 CP7666 CP7691 CP8014 CP8753 CP6681 CP7598 CP7681 CP7691I5 CP7696 CP7741 CP7771 CP8511 CP8751 CP8761
CAAGCP7595EA 1263.16 CAD
CAAGCP7595 CAAGCP7596 CAAGCP7666 CAAGCP7691 CAAGCP8014 CAAGCP8753 CAAGCP6681 CAAGCP7598 CAAGCP7681 CAAGCP7691I5 CAAGCP7696 CAAGCP7741 CAAGCP7771 CAAGCP8511 CAAGCP8751 CAAGCP8761
J&W CP-Sil 8 CB Columns, Agilent Technologies
Chromatography Columns
A general-purpose (5% phenyl) methlypolysiloxane phase that is excellent for method development.
  • (5% phenyl) methylpolysiloxane
  • Nonpolar, general-purpose phase
  • Bonded, crosslinked, solvent rinsable, and low bleed
  • High column-to-column reproducibility
  • Wide range of available dimensions
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-5, Rtx-5, BP-5, OV-5, 007-2(MPS-5), SE-52, SE-54, XTI-5, PTE-5, ZB-5, AT-5
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Low-polarity stationary phase