J&W CP-Select 624 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7414 CP7416 CP7412 CP7413 CP7415 CP7417
CACP7414EA 1317.7 CAD
CACP7414 CAAGCP7416 CAAGCP7412 CAAGCP7413 CAAGCP7415 CAAGCP7417
J&W CP-Select 624 CB Columns, Agilent Technologies
Chromatography Columns
A highly reproducible version of the popular 624 phase, with two to three times lower bleed than conventional columns.
  • Guaranteed for EPA volatiles with methods 524.2, 624 and 8015 for maximum confidence
  • Specified by the European Pharmacopoeia method V.3.3.9 for residual solvent analysis, providing compliancy
  • Excellent column-to-column reproducibility
  • Low bleed
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Select 624 CB, a 6% cyanopropyl, 94% dimethylpolysiloxane phase, is a highly reproducible version of the popular 624 phase and has two to three times lower bleed than conventional columns (bleed specification for a 30 m, 0.53 mm, df = 3.00 µm is 9 pA). It is synthesized and fully characterized to ensure maximum column-to-column reproducibility.