J&W CP-Chirasil-Dex CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7502 CP7502I5 CP7503
CAAGCP7502EA 1742.48 CAD
CAAGCP7502 CAAGCP7502I5 CAAGCP7503
J&W CP-Chirasil-Dex CB Columns, Agilent Technologies
Chromatography Columns
Delivers the highest resolution between isomers with guaranteed enantioselective stability for improved lifetime.
  • Cyclodextrin bonded to dimethylpolysiloxane for homogeneous enantioselectivity throughout the column
  • High resolution of isomers across a broad application range
  • Chemically bonded phase for excellent longevity
  • No need for derivatization, which improves productivity
  • Low elution temperature of polar compounds
  • Suitable for all injection techniques
  • Similar to LIPODEX C, Rt-β DEXm, β-DEX 110, β-DEX 120
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Chirasil-Dex CB consists of cyclodextrin directly bonded to dimethylpolysiloxane. This prevents the cyclodextrin from migrating to different locations in the surface film, delivering homogeneous enantioselectivity throughout the phase, which provides the highest resolution of isomers. It also guarantees stability of enantioselectivity; as a result, the lifetime of β-cyclodextrin capillary columns is significantly improved. CP-Chirasil-Dex CB permits low elution temperatures of polar compounds and is suitable for all injection techniques.