J&W CP-Al2O3/Na2SO4 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7565 CP7568 CP7565I5 CP7586
CAAGCP7565EA 2327.62 CAD
CAAGCP7565 CAAGCP7568 CAAGCP7565I5 CAAGCP7586
J&W CP-Al2O3/Na2SO4 Columns, Agilent Technologies
Chromatography Columns
Has a polar surface to separate ppm levels of C1 to C5 hydrocarbons in a main stream of C1 to C5 hydrocarbons.
  • Aluminum oxide PLOT columns offer high selectivity for separating ppm levels of C1 to C5 hydrocarbons in process streams
  • High-capacity thick films
  • No need for sub-ambient cooling simplifies operation
  • Choice of two selectivities covers a broad range of applications
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Al₂O₃/Na₂SO₄, Rt-Alumina PLOT, Alumina PLOT, Rt-Alumina BOND/Na₂SO₄, MXT-AluminaBOND/Na₂SO₄, Alumina sulfate PLOT
Agilent J&W CP-Al₂O₃/Na₂SO₄ is an aluminum oxide PLOT column that offers high selectivity for separating ppm levels of C1 to C5 hydrocarbons in a main stream of C1 to C5 hydrocarbons. The Na₂SO₄ deactivation salt provides a polar Al₂O₃ surface, so that unsaturated compounds such as ethylene and acetylene are retained longer. These columns analyze more compounds in a single run than packed columns, while still delivering higher resolution and faster analysis times. When compared to liquid stationary phases, CP-Al₂O₃/Na₂SO₄ provides increased selectivity and allows all C1 to C5 hydrocarbon isomers to be separated. These columns operate without the need for sub-ambient cooling.
Caution: NOTE: The KCl deactivation salt results in a relatively apolar Al₂O₃ surface while the Na₂SO₄ deactivation provides a polar surfaceUnsaturated compounds such as ethylene and acetylene (ethyne) are retained longer.