CrossLab CAM Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
112-2132 113-2132
CA112-2132EA 1253.9 CAD
CA112-2132 CA113-2132
CrossLab CAM Columns, Agilent Technologies
Chromatography Columns
A base-deactivated PEG column specifically designed for amine analysis, delivering excellent peak shape for primary amines.
  • Base-deactivated polyethylene glycol
  • Specifically designed for amines analysis
  • Excellent peak shape for primary amines
  • Replaces HP-Basicwax
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each GC column
  • Not bonded or crosslinked, not recommended for solvent rinsing
Agilent J&W CAM is a polyethylene glycol column that delivers excellent peak shape for primary amines. CAM replaces HP-BasicWax. The column is not bonded or crosslinked, so solvent rinsing is not recommended.