Agilent Bio SEC-5, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-2536 5190-2518 5190-2523 5190-2526 5190-2527 5190-2528 5190-2529 5190-2535 5190-2540 5190-2544 5190-2545 5190-6860 5190-6861 5190-6862
CAAG5190-2536EA 3296.43 CAD
CAAG5190-2536 CAAG5190-2518 CAAG5190-2523 CAAG5190-2526 CAAG5190-2527 CAAG5190-2528 CAAG5190-2529 CAAG5190-2535 CAAG5190-2540 CAAG5190-2544 CAAG5190-2545 CAAG5190-6860 CAAG5190-6861 CAAG5190-6862
Agilent Bio SEC-5, Agilent Technologies
Chromatography Columns
Widest pore size range SEC columns, from 100 to 2000 Å, with low operating pressures.
  • Maximum recovery for a broad range of size-based, biomolecule separations
  • Outstanding reproducibility and column lifetime
  • Excellent stability, even under high pH, high salt, and low salt conditions
  • Compatible with most aqueous buffers
Agilent Bio SEC-5 HPLC columns are packed with 5 μm silica particles coated with a proprietary, neutral, hydrophilic layer for maximum efficiency and stability. Our specially designed packing also promotes high pore volume, improvingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. pressure 240 bar
pH range 2 - 8.5
Temperature limit 80 °C
Wetted materials Phosphate Buffer