Diamond® APEX™ Pipettors, Single-Channel, Globe Scientific
Supplier: Globe Scientific
3351-1 3351-10 3351-20 3351-50 3351-100 3351-200 3351-1000 3351-5000 3351-10000 3351-COMBO 3351-COMBOFT 3352-2 3352-5 3352-10 3352-20 3352-25 3352-50 3352-100 3352-200 3352-250 3352-500 3352-1000 3352-2000 3352-5000 3352-10000
76509-178EA 468.6 CAD
76509-178 76509-180 76509-182 76509-184 76509-186 76509-188 76509-190 76509-192 76509-194 76509-196 76509-198 76509-202 76509-204 76509-206 76509-208 76509-210 76509-212 76509-214 76509-216 76509-218 76509-220 76509-222 76509-224 76509-226 76509-228
Diamond® APEX™ Pipettors, Single-Channel, Globe Scientific
Pipettes Micropipettes
The Diamond® APEX™ series of pipettors are quite simply one of the best lines of pipettors available today.
  • Fully autoclavable and UV resistant
  • Three-year warranty
  • Color coded plungers for easy volume identification
  • Virtually fatigue-free pipetting with low plunger force
  • Magnet assisted piston for extremely high accuracy and consistency
  • Ergonomic design
  • Pipettors feature locking mechanism to prevent accidental volume changes
The magnetic assisted piston ensures the best consistency with each plunger motion to the first and second stops for aspiration,More Product Information