J&W VF-5ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP8946 CP8962 CP9034 CP8944 CP8939 CP8945 CP8957 CP8960 CP9043
CAAGCP8946EA 1250.9 CAD
CAAGCP8946 CAAGCP8962 CAAGCP9034 CACP8944 CAAGCP8939 CAAGCP8945 CAAGCP8957 CAAGCP8960 CAAGCP9043
J&W VF-5ms Columns, Agilent Technologies
Chromatography Columns
An inert, low-bleed 5% phenylmethyl polysiloxane phase for increased sensitivity, accuracy, and instrument uptime for semipolar and polar compounds.
  • Highly inert 5% phenylmethyl polysiloxane column for increased sensitivity, accuracy, and instrument uptime
  • Minimal column bleed improves sensitivity (ultra-low bleed specification of 1 pA @ 325 °C (for 30000×0.25 mm, 0.25 μm))
  • Slightly higher polarity than VF-1ms for improved selectivity for aromatic compounds
  • Excellent selectivity and inertness, applicable to a wide range of semipolar and even polar compounds
  • 0.15 mm id columns available for high efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-5ms is an inert 5% phenylmethyl polysiloxane column that delivers increased sensitivity, accuracy, and instrument uptime. The column has the lowest guaranteed bleed specification of 1 pA at 325 °C (for 30 m, 0.25 mm, 0.25 µm). This column has slightly higher polarity than VF-1ms, resulting in better selectivity for aromatic compounds. This selectivity, combined with superior inertness, makes these columns applicable to a wide range of semipolar and even polar compounds, such as phenols.