PolarGel, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1117-6830 PL1117-1800 PL1117-1830
CAAGPL11176830EA 3628.68 CAD
CAAGPL11176830 CAAGPL11171800 CAAGPL11171830
PolarGel, Agilent Technologies
Chromatography Columns
PolarGel GPC columns are designed for use with polar organic solvents and water/organic mixtures.
  • Proprietary surface chemistry is compatible with a wide range of polar solvents, modifiers, and mixtures
  • Excellent performance for polymers which dissolve in DMF, DMSO, NMP, DMAc, and THF/water mixtures
PolarGel GPC columns are designed for use with polar organic solvents such as DMF, DMSO, NMP, and DMAc, as well as water/organic mixtures. These eluents are very useful in GPC/SEC to separate polar polymers, such as resins, starches, acrylics, biopolymers,More Product Information