PL aquagel-OH Analytical, Agilent Technologies

Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1120-6520 PL1120-6830 PL1149-6260 PL1149-6820 PL1149-6850 PL1149-6860 PL1149-6240 PL1149-6801 PL1549-5800
CAPL1120-6520EA 3449.96 CAD
CAPL1120-6520 CAPL1120-6830 CAAGPL11496260 CAAGPL11496820 CAAGPL11496850 CAAGPL11496860 CAAGPL11496240 CAAGPL11496801 CAAGPL15495800
PL aquagel-OH Analytical, Agilent Technologies
Chromatography Columns
PL aquagel-OH SEC columns separate water-soluble polymers and large molecules.

  • Efficient and reliable size exclusion chromatography (SEC) column
  • Meets USP L25, tolerates high buffer concentrations, pH range 2 to 10, temperatures up to 90 °C and organic solvents (up to 50% methanol)
  • Various MW options provide tailored separations
  • Well-tested resistance against interaction with neutral, multiply charged, polyanionic and polycationic samples

The PL aquagel-OH size exclusion chromatography (SEC) column offers efficient and reliable More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 150 bar 150 bar
pH range 2-10 2-10
Temperature limit 90 °C 90 °C
Wetted materials 0.02% Sodium azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR