InfinityLab Poroshell 120 for NP/SFC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
693775-901 693675-901 693770-901 693975-301 693975-901 695675-301 695675-901 695770-901 695775-901T 695975-301 695975-901 699675-901 699775-901 699975-901 821725-926 821725-944 823750-926
CAAG693775-901EA 1656.18 CAD
CAAG693775-901 CAAG693675-901 CAAG693770-901 CAAG693975-301 CAAG693975-901 CAAG695675-301 CAAG695675-901 CAAG695770-901 CAAG695775901T CAAG695975-301 CAAG695975-901 CAAG699675-901 CAAG699775-901 CAAG699975-901 CAAG821725-926 CAAG821725-944 CAAG823750-926
InfinityLab Poroshell 120 for NP/SFC, Agilent Technologies
Chromatography Columns
Modern column technology based on superficially porous particles for normal phase LC separations.
  • Superficially porous, 'core-shell' particles that provide both improved throughput and higher resolution
  • A scalable family of particles—1.9 μm, 2.7 μm, and 4 μm—enables you to get the best from your methods and instruments
  • Tested in SFC so you can benefit from faster separations and sharper peaks and worry even less about system pressures
  • The InfinityLab Poroshell 120 HILIC has been highlighted as one of the most universal, best-in-class superficially porous SFC columns in independent research publications
  • Excellent lot-to-lot reproducibility for confidence in your separation results
  • Part of the InfinityLab family – designed for ease-of-use and maximum compatibility with Agilent instruments and supplies
  • UHPLC guard columns to further extend the life of your analytical column
  • Superior peak shape for faster, more accurate results due to high-purity silica and advanced bonding chemistries
  • Enhanced usability, traceability, and security with a preprogrammed ID tag
Agilent InfinityLab Poroshell 120 superficially porous columns for normal phase HPLC and supercritical fluid chromatography (SFC) offer exceptional efficiency and reliability. Unique, chemistries including Poroshell 120 EC-CN, HILIC, and HILIC-Z (HILIC phases in SFC only), are available in up to three particle diameters, 1.9 μm, 2.7 μm, and 4 μm. These options provide a broad range of selectivity for your analytical method development and are scalable from traditional HPLC to UHPLC and through to ultralow dispersion UHPLC.