InfinityLab PlusPore, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1113-6325 PL1113-1320 PL1113-1325 PL1513-1320 PL1513-5300 PL1513-5325 PL1513-5500 PL1513-5520 PL1913-5500
CAAGPL11136325EA 4177.9 CAD
CAAGPL11136325 CAAGPL11131320 CAAGPL11131325 CAAGPL15131320 CAAGPL15135300 CAAGPL15135325 CAAGPL15135500 CAAGPL15135520 CAAGPL19135500
InfinityLab PlusPore, Agilent Technologies
Chromatography Columns
PlusPore GPC columns offer the highest resolution gel permeation chromatography.
  • PlusPore GPC columns offer a major resolution improvement over traditional mixed medias and single pore columns
  • High-efficiency small particles use multipore technology, maximizing resolution without sacrificing molecular weight range or lifetime
  • OligoPore uses unique ultrahigh pore volume media to achieve superior separation of polymers below 3,300 Daltons
PLgel individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. ColumnsMore Product Information