J&W HP-PONA Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091S-001 19091S-001E
CA19091S-001EA 2374.96 CAD
CA19091S-001 CAAG19091S001E
J&W HP-PONA Columns, Agilent Technologies
Chromatography Columns
Specially configured for the analysis of petroleum process products, including paraffins, isoparaffins, aromatics, napthalenes, and olefins.
  • 100% Dimethylpolysiloxane
  • Configured for the analysis of petroleum process products
  • Tested to ensure the resolution of m-xylene from p-xylene and of cyclopentane from 2,3-dimethylbutane
  • PONA, PIANO
  • High resolution
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: Petrocol DH, SPB-1, 007-1, Rtx-1, MXT-1, Rtx-1PONA, Rtx-DHA
Agilent J&W HP-PONA columns are configured to deliver excellent, high-resolution performance for the detailed analysis of petroleum process products, including paraffins, isoparaffins, aromatics, napthalenes, and olefins (PIANO). These 100% dimethylpolysiloxane columns are bonded, crosslinked, and solvent rinsable, and are tested to strict industry QC standards during manufacture, and to ensure the resolution of m-xylene from p-xylene and of cyclopentane from 2,3-dimethylbutane.