HP-88 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
112-8867 112-88A7
CA112-8867EA 1857.23 CAD
CA112-8867 CAAG112-88A7
HP-88 Columns, Agilent Technologies
Chromatography Columns
High-polarity cyanopropylpolysiloxane column designed for the separation of cis/trans fatty acid methyl esters.
  • (88% Cyanopropy) aryl-polysiloxane
  • 250/260 °C upper temperature limits
  • High polarity
  • Designed for separation of cis/trans fatty acid methyl esters (FAMEs)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-88 is a high-polarity column designed for the separation of fatty acid methyl esters (FAMEs). It provides great resolution ofMore Product Information