HP-5ms Ultra Inert Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091S-436UI 19091S-431UI 19091S-433UI 19091S-133UI 19091S-413UI 19091S-433UIE G3903-61005 19091S-233UI 19091S-413UI-INT 19091S-430UI-INT
CA19091S-436UIEA 2309.45 CAD
CA19091S-436UI CA19091S-431UI CA19091S-433UI CAAG19091S133U CAAG19091S413U CAAG19091S433U CAAGG390361005 CAAG19091S233U CAAG19091S413N CAAG19091S430U
HP-5ms Ultra Inert Columns, Agilent Technologies
Chromatography Columns
A nonpolar (5%-phenyl)-methylpolysiloxane, low-bleed GC column for trace-level analysis of acidic and basic compounds.
  • Excellent for trace-level analysis of acids and bases
  • (5%-Phenyl)-methylpolysiloxane has identical selectivity to HP-5MS
  • Specifically tested for inertness for active compounds, including acidic and basic compounds
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Nonpolar with very low bleed characteristics, ideal for GC/MS
  • Bonded, crosslinked, and solvent rinsible
  • Equivalent to USP phase G27
  • Tested with the
More Product Information