J&W Ultra Inert HP-1ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091S-733UI 19091S-612UI 19091S-633UI 19091S-677UI 19091S-713UI 19091S-911UI-INT 19091S-911UI 19091S-913UI-INT 19091S-913UI 19091S-933UI-INT 19091S-933UI
CAAG19091S733UEA 1355.21 CAD
CAAG19091S733U CAAG19091S612U CAAG19091S633U CAAG19091S677U CAAG19091S713U CAAG19091S911I CAAG19091S911U CAAG19091S913I CAAG19091S913U CAAG19091S933I CAAG19091S933U
J&W Ultra Inert HP-1ms Columns, Agilent Technologies
Chromatography Columns
Nonpolar PDMS with outstanding inertness that delivers excellent peak shapes for challenging active analytes.
  • Individually tested with Ultra Inert 1 ms test probe mixture
  • Consistent column inertness performance
  • Exceptionally low column bleed
  • Great peak shapes for challenging active analytes
  • Excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm id column configuration for higher sample throughput
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Low-polarity stationary phase