J&W HP-1ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19091S-913 19091S-931 19091S-933 19091S-602 19091S-936 19091S-612 19091S-633 19091S-677 19091S-713 19091S-733 19091S-833 19091S-916 19091S-733E 19091S-913E 19091S-936-INT
CA19091S-913EA 1354.34 CAD
CA19091S-913 CA19091S-931 CA19091S-933 CA19091S-602 CA19091S-936 CAAG19091S-612 CAAG19091S-633 CAAG19091S-677 CAAG19091S-713 CAAG19091S-733 CAAG19091S-833 CAAG19091S-916 CAAG19091S733E CAAG19091S913E CAAG19091S936I
J&W HP-1ms Columns, Agilent Technologies
Chromatography Columns
An excellent nonpolar, general-purpose column for analysis of hydrocarbons, pesticides, PCBs, sulfur compounds, and more.
  • 100% Dimethylpolysiloxane
  • Identical selectivity to HP-1
  • Nonpolar
  • Low bleed characteristics
  • Excellent general-purpose column
  • Improved signal-to-noise ratio for better sensitivity and mass spectral integrity
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W HP-1ms is a high-quality, 100More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Low-polarity stationary phase