Hi-Plex, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1170-2820 PL1570-5810 PL1170-1850 PL1170-2823 PL1171-6140 PL1570-6810 PL1F70-6830 PL1F70-6850
CAAGPL11702820EA 2242.36 CAD
CAAGPL11702820 CAAGPL15705810 CAAGPL11701850 CAAGPL11702823 CAAGPL11716140 CAAGPL15706810 CAAGPL1F706830 CAAGPL1F706850
Hi-Plex, Agilent Technologies
Chromatography Columns
Hi-Plex HPLC columns for analysis of carbohydrates, sugars, and organic acids.
  • Agilent's recommended column for carbohydrate analysis
  • Operates on a standard HPLC and does not require specialized equipment
  • Simple operating conditions: water or dilute acid, isocratic conditions, up to 30 % acetonitrile as a modifier
  • Available in 8 μm and 10 μm particle sizes
  • Complies with USP L17, USP L19, and USP L34
Agilent Hi-Plex HPLC columns provide reliable carbohydrate analysis with simple instrument requirements. Unlike ion chromatography columns, Hi-Plex columns use pure water or dilute acid on a standard HPLC, and do not require ion regeneration or suppressors. The media consists of robust a sulfonated cross-linked styrene-divinylbenzene gel in hydrogen form or is impregnated with various metal cations with different selectivity, as described in USP L17, L19, L34, and L58.