GS-CarbonPLOT Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
113-3133 115-3133
CA113-3133EA 1381.73 CAD
CA113-3133 CA115-3133
GS-CarbonPLOT Columns, Agilent Technologies
Chromatography Columns
A high-stability, carbon-layer stationary phase with unique selectivity for inorganic and organic gases.
  • High-stability, carbon-layer stationary phase
  • Unique selectivity for inorganic and organic gases
  • Extended temperature limit of 360 °C
  • Ideal for GC/MS, due to no particle generation
  • Retention stability not affected by water
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W GS-CarbonPLOT uses a high-stability, carbon-layer stationary phase toMore Product Information