J&W EZ-Guard Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP9010 CP9013 CP9011 CP9012 CP9014 CP9021 CP9023
CAAGCP9010EA 1415.98 CAD
CAAGCP9010 CAAGCP9013 CAAGCP9011 CAAGCP9012 CAAGCP9014 CAAGCP9021 CAAGCP9023
J&W EZ-Guard Columns, Agilent Technologies
Chromatography Columns
Combines a VF column with a built-in guard column to avoid problems associated with unions.
  • Built-in guard columns eliminate the need for press-fit connectors
  • Minimizes front-end contamination and increases column lifetime
  • Improves focusing sample onto the front of the column for better peak shape
  • Minimizes MSD contamination originating from the column (when used as transfer line to the MS detector)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
EZ-Guard columns combine an Agilent J&W VF column with a built-in guard column. The first 5 or 10 m section of the column (depending on the configuration) is not coated with stationary phase, but has been deactivated. Eliminating the column connection between the guard and analytical sections ensures 100% leak-free performance.